missing translation for 'onlineSavingsMsg'
Learn More

1-Heptyne, TRC

Product Code. 30392005
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392005 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392005 Supplier Toronto Research Chemicals Supplier No. TRCH28820050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Heptyne
CAS 628-71-7
Molecular Formula C7H12
Quantity 50 g
Synonym 1-n-Heptyne, Amylacetylene, NSC 9710, Pentylacetylene
InChI Formula InChI=1S/C7H12/c1-3-5-7-6-4-2/h1H,4-7H2,2H3
SMILES CCCCCC#C
IUPAC Name hept-1-yne
Molecular Weight (g/mol) 96.17
Formula Weight 96.0939
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.