missing translation for 'onlineSavingsMsg'
Learn More

1-Heptyne, 99%

Product Code. p-140805 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10442611

Brand: Thermo Scientific Acros 223460250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-71-7 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-71-7
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00009529
InChI Key YVXHZKKCZYLQOP-UHFFFAOYSA-N
Synonym 1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, acmc-1aw2o
PubChem CID 12350
IUPAC Name hept-1-yne
SMILES CCCCCC#C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -81°C
Color Colorless to Green-Yellow
Density 0.7300g/mL
Boiling Point 99°C to 100°C
Flash Point -002°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)4C∽CH
Refractive Index 1.4070 to 1.409
Quantity 25 mL
Beilstein 01, 256
Specific Gravity 0.73
Formula Weight 96.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Heptyne, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Heptyne
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.