missing translation for 'onlineSavingsMsg'
Learn More

1-Heptyne, 99%

Product Code. 11489023
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489023 25 g 25g
11474170 5 g 5g
11499023 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489023 Supplier Thermo Scientific Alfa Aesar Supplier No. A11130.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-71-7 | C7H12 | 96.173 g/mol

1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-71-7
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00009529
InChI Key YVXHZKKCZYLQOP-UHFFFAOYSA-N
Synonym 1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, acmc-1aw2o
PubChem CID 12350
IUPAC Name hept-1-yne
SMILES CCCCCC#C

Specifications

Melting Point -81°C
Density 0.736
Boiling Point 98°C to 100°C
Flash Point −10°C (14°F)
Odor Unpleasant
Refractive Index 1.4095
Quantity 25 g
UN Number UN3295
Beilstein 1733518
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 96.17
Percent Purity 99%
Chemical Name or Material 1-Heptyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.