missing translation for 'onlineSavingsMsg'
Learn More

1-Heptylamine, 98+%

Product Code. 11429046
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429046 25 g 25g
11408750 50 g 50g
11418750 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429046 Supplier Thermo Scientific Alfa Aesar Supplier No. A17922.14
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-68-2 | C7H17N | 115.22 g/mol

1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

Solubility
Soluble in water 6791 mg/L @ 25°C.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-68-2
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008244
InChI Key WJYIASZWHGOTOU-UHFFFAOYSA-N
Synonym heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
PubChem CID 8127
IUPAC Name heptan-1-amine
SMILES CCCCCCCN

Specifications

Melting Point -23°C
Density 0.775
Boiling Point 154°C to 156°C
Flash Point 35°C (95°F)
Odor Amine-like
Linear Formula CH3(CH2)6NH2
Refractive Index 1.4245
Quantity 25 g
UN Number UN2734
Beilstein 1731688
Sensitivity Air Sensitive
Solubility Information Soluble in water 6791mg/L @ 25°C.
Formula Weight 115.22
Percent Purity ≥98%
Chemical Name or Material 1-Heptylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.