missing translation for 'onlineSavingsMsg'
Learn More

1-Hepten-3-ol, 98%

Product Code. 11469110
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469110 10 g 10g
11479110 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469110 Supplier Thermo Scientific Alfa Aesar Supplier No. A14651.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4938-52-7 | C7H14O | 114.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4938-52-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00021940
InChI Key PZKFYTOLVRCMOA-UHFFFAOYSA-N
Synonym 1-hepten-3-ol, heptene-1-ol-3, butyl vinyl carbinol, acmc-1aoyf, 1-hepten-3-ol, analytical standard, 1-hepten-3-ol, fg, 1-hepten-3-ol, purum gc
PubChem CID 21057
IUPAC Name hept-1-en-3-ol
SMILES CCCCC(C=C)O

Specifications

Density 0.836
Boiling Point 154°C to 156°C
Flash Point 54°C (129°F)
Refractive Index 1.433
Quantity 10 g
UN Number UN1987
Beilstein 1720340
Formula Weight 114.18
Percent Purity 98%
Chemical Name or Material 1-Hepten-3-ol
compliance-icons
Product Identifier
  • 1-Hepten-3-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 1-Hepten-3-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.