missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, TRC

Product Code. 30396632
Change view
Click to view available options
Quantity:
5 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396632 5 mL
30406578 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396632 Supplier Toronto Research Chemicals Supplier No. TRCH2811955ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7 H16 O
Molecular Weight (g/mol) 116.2
Synonym 1-Heptanol (9CI, ACI), Heptyl alcohol (8CI), 1-Hydroxyheptane, Enanthic alcohol, Gentanol, n-Heptan-1-ol, n-Heptanol, n-Heptyl alcohol, NSC 3703
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Quantity 5 mL
InChI Formula InChI=1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3
Formula Weight 116.1201
Chemical Name or Material 1-Heptanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.