missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, 99%

Product Code. 11405153
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405153 2500 mL 2500mL
11485143 100 mL 100mL
11495143 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405153 Supplier Thermo Scientific Alfa Aesar Supplier No. A12793.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-70-6 | C7H16O | 116.20 g/mol

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Melting Point -34°C
Density 0.82
Boiling Point 175°C to 176°C
Flash Point 73°C (163°F)
Assay Percent Range 99%
Odor Characteristic
Linear Formula CH3(CH2)6OH
Refractive Index 1.424
Quantity 2500 mL
Beilstein 1731686
Merck Index 14,4661
Solubility Information Soluble in water 1.7g/L at 25°C.
Formula Weight 116.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Heptanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.