missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanethiol, 98%

Product Code. 11377126
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377126 100 g 100g
11367126 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377126 Supplier Thermo Scientific Alfa Aesar Supplier No. L07079.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1639-09-4 | C7H16S | 132.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1639-09-4
Molecular Formula C7H16S
Molecular Weight (g/mol) 132.265
MDL Number MFCD00004911
InChI Key VPIAKHNXCOTPAY-UHFFFAOYSA-N
Synonym 1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
PubChem CID 15422
IUPAC Name heptane-1-thiol
SMILES CCCCCCCS

Specifications

Melting Point -43°C
Density 0.854
Boiling Point 173°C to 175°C
Flash Point 46°C (114°F)
Refractive Index 1.4515
Quantity 100 g
UN Number UN3336
Beilstein 1731687
Sensitivity Air Sensitive
Formula Weight 132.27
Percent Purity 98%
Chemical Name or Material 1-Heptanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.