missing translation for 'onlineSavingsMsg'
Learn More

1-Heptane Sulfonic Acid Sodium Salt, Ion Pair Chromatography, Solution 0.1M, Fisher Chemical

Product Code. 10244480 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Amber glass bottle
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244480 250 mL Amber glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244480 Supplier Fisher Chemical Supplier No. H/0166/07

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 22767-50-6
Color Colorless
Physical Form Liquid
Quantity 250 mL
Molecular Formula C7H15NaO3S
MDL Number 7543
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;heptane-1-sulfonate
Molecular Weight (g/mol) 202.244
PubChem CID 23672332
Packaging Amber glass bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.