missing translation for 'onlineSavingsMsg'
Learn More

1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™

Product Code. p-7073049 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
100 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
100g
25g
5kg
This item is not returnable. View return policy

Product Code. 10315850

Brand: Fisher Chemical H/0168/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical, Quantity: 100g, Packaging: Plastic powder jar, CAS: 22767-50-6, White, Melting Point: 300 deg.C, Molecular Formula: C7H15NaO3S, Molecular Weight: 202.244

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Molecular Weight (g/mol) 202.244
MDL Number 7543
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
PubChem CID 23672332
IUPAC Name sodium;heptane-1-sulfonate
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White
Physical Form Powder/Solid
Quantity 25 g
Formula Weight 202.25g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • 1-Heptanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.