missing translation for 'onlineSavingsMsg'
Learn More

1-(Heptafluorobutyryl)imidazole, TRC

Product Code. 30401345
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401345 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401345 Supplier Toronto Research Chemicals Supplier No. TRCH2810602.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Heptafluorobutyryl)imidazole
CAS 32477-35-3
Molecular Formula C7 H3 F7 N2 O
Quantity 2.5 g
Synonym 2,2,3,3,4,4,4-Heptafluoro-1-(1H-imidazol-1-yl)-1-butanone, 1-(2,2,3,3,4,4,4-Heptafluoro-1-oxobutyl)-1H-imidazole, NSC 151966
InChI Formula InChI=1S/C7H3F7N2O/c8-5(9,6(10,11)7(12,13)14)4(17)16-2-1-15-3-16/h1-3H
SMILES FC(F)(F)C(F)(F)C(F)(F)C(=O)n1ccnc1
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one
Molecular Weight (g/mol) 264.1
Formula Weight 264.0134
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.