missing translation for 'onlineSavingsMsg'
Learn More

1-Heptadecene, 97%, Thermo Scientific™

Product Code. 10642382 Shop All Thermo Scientific Products
Click to view available options
:
5mL; Glass bottle
This item is not returnable. View return policy

Product Code. 10642382

Brand: Thermo Scientific 338330050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6765-39-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 10°C
Color Yellow
Density 0.7900g/mL
Boiling Point 300°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C17H34
Linear Formula CH2CH(CH2)14CH3
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
MDL Number MFCD00009000
Quantity 5 mL
Specific Gravity 0.79
Synonym 1-heptadecene, heptadecene, hexahydroaplotaxene, alkenes, c>10 alpha, heptadecen, alkenes, c>10 .alpha., alpha-olefin wax, alpha olefins petroleum , c10 cut, c>10 alpha-alkenes, heptadecene related
InChI Key ADOBXTDBFNCOBN-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC=C
IUPAC Name heptadec-1-ene
Molecular Weight (g/mol) 238.46
PubChem CID 23217
ChEBI CHEBI:64502
Formula Weight 238.46
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Heptadecene, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.