missing translation for 'onlineSavingsMsg'
Learn More

1-Heptadecanol, 97%

Product Code. 10216900 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10216900

Brand: Thermo Scientific Acros 221230250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1454-85-9 | C17H36O | 256.474 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454-85-9
InChI Key GOQYKNQRPGWPLP-UHFFFAOYSA-N
Synonym 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1
PubChem CID 15076
ChEBI CHEBI:77470
IUPAC Name heptadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCO

Specifications

Melting Point 51.0°C to 56.0°C
Color White
Boiling Point 308.5°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C17H36O
Linear Formula CH3(CH2)16OH
Quantity 25 g
Solubility Information Solubility in water: slightly soluble in water.
Molecular Weight (g/mol) 256.474
Formula Weight 256.46
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Heptadecanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.