missing translation for 'onlineSavingsMsg'
Learn More

1-Formyl Mestanolone, TRC

Product Code. 30407726
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407726 1 mg 1mg
30408432 25 mg 25mg
30397853 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407726 Supplier Toronto Research Chemicals Supplier No. TRCF7006851MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Formyl Mestanolone
Analyte or Component Names 1-Formyl Mestanolone
Shipping Condition Room Temperature
Molecular Formula C21H32O3
Synonym (1S,5S,8R,9S,10S,13S,14S,17S)-17-Hydroxy-10,13,17-trimethyl-3-oxohexadecahydro-1H-cyclopenta[a]phenanthrene-1-carbaldehyde
InChI Formula InChI=1S/C21H32O3/c1-19-8-6-18-16(17(19)7-9-20(19,2)24)5-4-13-10-15(23)11-14(12-22)21(13,18)3/h12-14,16-18,24H,4-11H2,1-3H3/t13-,14?,16-,17-,18-,19-,20-,21+/m0/s1
Solution Type Neat
SMILES [H][C@]12CC(CC(C([H])=O)[C@]1(C)[C@]3([H])[C@@]([C@@](CC[C@@]4(O)C)([H])[C@]4(C)CC3)([H])CC2)=O
Industry Type Forensics
Molecular Weight (g/mol) 332.48
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.