missing translation for 'onlineSavingsMsg'
Learn More

1-Fmoc-azetidine-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 15473608
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473608 250 mg 250mg
15483608 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473608 Supplier Thermo Scientific Alfa Aesar Supplier No. H57059.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193693-64-0
Molecular Formula C19H17NO4
Molecular Weight (g/mol) 323.348
MDL Number MFCD01321018
InChI Key QDEJCUHGSHSYQH-UHFFFAOYSA-N
Synonym 1-fmoc-azetidine-3-carboxylic acid, fmoc-azetidine-3-carboxylic acid, fmoc-l-azetidine-3-carboxylic acid, 1-9h-fluoren-9-yl methoxy carbonyl azetidine-3-carboxylic acid, 1-9h-fluoren-9-ylmethoxycarbonyl azetidine-3-carboxylic acid, 1-fmoc-azetidine-3-carboxylicacid, 1-9h-fluoren-9-ylmethoxy carbonyl azetidine-3-carboxylic acid, acmc-209ex6, ksc541g2b
PubChem CID 2756148
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonyl)azetidine-3-carboxylic acid
SMILES C1C(CN1C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Density 1.368
Boiling Point 540°C to 545°C
Flash Point 282°C (540°F)
Quantity 250 mg
Formula Weight 323.34
Percent Purity 95%
Chemical Name or Material 1-Fmoc-azetidine-3-carboxylic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.