missing translation for 'onlineSavingsMsg'
Learn More

1-Fmoc-3-(5-methyl-1,2,4-oxadiazol-3-yl)indole, 97%, Thermo Scientific™

Product Code. 15484446
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858240-59-1
Molecular Formula C26H19N3O3
Molecular Weight (g/mol) 421.456
MDL Number MFCD22988970
InChI Key KPOAIVGPPNHNQJ-UHFFFAOYSA-N
Synonym 9h-fluoren-9-ylmethyl 3-5-methyl-1,2,4-oxadiazol-3-yl indole-1-carboxylate, 1-fmoc-3-5-methyl-1,2,4-oxadiazol-3-yl indole
PubChem CID 98000075
IUPAC Name 9H-fluoren-9-ylmethyl 3-(5-methyl-1,2,4-oxadiazol-3-yl)indole-1-carboxylate
SMILES CC1=NC(=NO1)C2=CN(C3=CC=CC=C32)C(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Quantity 1 g
Formula Weight 421.45
Percent Purity 97%
Chemical Name or Material 1-Fmoc-3-(5-methyl-1,2,4-oxadiazol-3-yl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'