missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-6-methoxy-naphthalene, TRC

Product Code. 30400861
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400861 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400861 Supplier Toronto Research Chemicals Supplier No. TRCF5933601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-fluoro-6-methoxynaphthalene
CAS 853192-64-0
Molecular Formula C11 H9 F O
Quantity 1 g
InChI Formula InChI=1S/C11H9FO/c1-13-9-5-6-10-8(7-9)3-2-4-11(10)12/h2-7H,1H3
SMILES COc1ccc2c(F)cccc2c1
IUPAC Name 1-fluoro-6-methoxynaphthalene
Molecular Weight (g/mol) 176.187
Formula Weight 176.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.