missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-4-nitrobenzene, 99%

Product Code. 11488903
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488903 2500 g 2500g
11468903 100 g 100g
11478903 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488903 Supplier Thermo Scientific Alfa Aesar Supplier No. A11057.0E
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 350-46-9 | C6H4FNO2 | 141.101 g/mol

1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-46-9
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD00007282
InChI Key WFQDTOYDVUWQMS-UHFFFAOYSA-N
Synonym 4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene
PubChem CID 9590
IUPAC Name 1-fluoro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])F

Specifications

Melting Point 22°C to 24°C
Density 1.33
Boiling Point 204°C to 206°C
Flash Point 83°C (181°F)
Refractive Index 1.5315
Quantity 2500 g
UN Number UN2811
Beilstein 606923
Solubility Information Insoluble in water.
Formula Weight 141.1
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.