missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-iodobenzene, 99%

Product Code. 11488970
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488970 100 g 100g
11478970 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488970 Supplier Thermo Scientific Alfa Aesar Supplier No. A11706.22
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1121-86-4 | C6H4FI | 222.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-86-4
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001044
InChI Key VSKSBSORLCDRHS-UHFFFAOYSA-N
Synonym 3-fluoroiodobenzene, m-fluoroiodobenzene, 3-iodofluorobenzene, m-iodofluorobenzene, benzene, 1-fluoro-3-iodo, m-fluorobenzene, 1-fluoro-3-iodo-benzene, 1-iodo-3-fluorobenzene, 3-fluoro-1-iodobenzene, m-fluorjodbenzol
PubChem CID 70725
IUPAC Name 1-fluoro-3-iodobenzene
SMILES FC1=CC=CC(I)=C1

Specifications

Melting Point -42°C
Density 1.898
Boiling Point 77°C to 78°C (19 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.584
Quantity 100 g
Beilstein 2039305
Sensitivity Light sensitive
Formula Weight 222
Percent Purity 99%
Chemical Name or Material 1-Fluoro-3-iodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.