missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-iodobenzene, 99%, stabilized

Product Code. 10134460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10134460 25 g Glass bottle 25g
10246690 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10134460 Supplier Thermo Scientific Acros Supplier No. 204480250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1121-86-4 | C6H4FI | 222.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-86-4
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001044
InChI Key VSKSBSORLCDRHS-UHFFFAOYSA-N
Synonym 3-fluoroiodobenzene, m-fluoroiodobenzene, 3-iodofluorobenzene, m-iodofluorobenzene, benzene, 1-fluoro-3-iodo, m-fluorobenzene, 1-fluoro-3-iodo-benzene, 1-iodo-3-fluorobenzene, 3-fluoro-1-iodobenzene, m-fluorjodbenzol
PubChem CID 70725
IUPAC Name 1-fluoro-3-iodobenzene
SMILES FC1=CC=CC(I)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow or Pink
Density 1.8900g/mL
Boiling Point 77°C to 78°C (19.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4I
Refractive Index 1.5830 to 1.585
Quantity 25 g
Beilstein 05, III, 578
Specific Gravity 1.89
Formula Weight 222
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-3-iodobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Fluoro-3-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.