missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-iodo-5-nitrobenzene, 98%, Thermo Scientific™

Product Code. 10621115 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10621115 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10621115 Supplier Thermo Scientific Acros Supplier No. 378500010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3819-88-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 77°C to 79°C
Color Brown or Khaki to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H3FINO2
Linear Formula NO2C6H3I(F)
MDL Number MFCD00007219
Quantity 1 g
Synonym 3-fluoro-5-iodonitrobenzene, benzene, 1-fluoro-3-iodo-5-nitro, acmc-1cr1w, 5-fluoro-3-nitroiodobenzene, ksc495i2h, 1-iodo-3-fluoro-5-nitrobenzene, 1-fluoro-3-iodo-5-nitro-benzene, 1-fluoro-3-iodo-5-nitrobenzene
InChI Key MXPYCSFCKXSPAB-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1F)I)[N+](=O)[O-]
IUPAC Name 1-fluoro-3-iodo-5-nitrobenzene
Molecular Weight (g/mol) 266.99
PubChem CID 259086
Formula Weight 266.99
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 1-Fluoro-3-iodo-5-nitrobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Fluoro-3-iodo-5-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.