missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
Solubility
Immiscible in water.
Notes
Avoid excessive heat and light.
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
Solubility
Immiscible in water.
Notes
Avoid excessive heat and light.
Specifications
Melting Point | °C to 8°C |
Density | 1.340 |
Boiling Point | 215°C |
Flash Point | 94°C (201°F) |
Assay Percent Range | 99% |
Refractive Index | 1.5317 |
Quantity | 50 g |
UN Number | UN2810 |
Beilstein | 607262 |
Solubility Information | Immiscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'