missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynylcyclopentanol, 98+%
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
Descripción
1-Ethynylcyclopentanol is used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethynylcyclopentanol is used in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
1-Ethynylcyclopentanol is used in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Especificaciones
Especificaciones
| Melting Point | 23°C to 27°C |
| Density | 0.962 |
| Boiling Point | 156°C to 159°C |
| Flash Point | 48°C (118°F) |
| Odor | Irritating |
| Refractive Index | 1.474 |
| Quantity | 5 g |
| UN Number | UN1325 |
| Beilstein | 1924167 |
| Solubility Information | Insoluble in water. |
| Mostrar más |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido