missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylcyclohexanol, 99%

Product Code. 11323256
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323256 100 g 100g
11333256 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323256 Supplier Thermo Scientific Alfa Aesar Supplier No. L12591.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-27-3 | C8H12O | 124.183 g/mol

1-Ethynylcyclohexanol is used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylcyclohexanol is used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (10g/L at 20°C), diethyl ether, alcohols, esters, and ketone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-27-3
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00003858
InChI Key QYLFHLNFIHBCPR-UHFFFAOYSA-N
Synonym 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol
PubChem CID 6525
IUPAC Name 1-ethynylcyclohexan-1-ol
SMILES C#CC1(CCCCC1)O

Specifications

Melting Point 30°C to 33°C
Density 0.973
Boiling Point 180°C
Flash Point 73°C (163°F)
Odor Mild
Refractive Index 1.483
Quantity 100 g
UN Number UN2811
Beilstein 471404
Solubility Information Soluble in water (10g/L at 20°C),diethyl ether,alcohols,esters,and ketone.
Formula Weight 124.18
Percent Purity 99%
Chemical Name or Material 1-Ethynylcyclohexanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.