missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-fluorobenzene, 98%

Product Code. 10364742 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10364742 1 g Glass bottle 1g
11934531 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10364742 Supplier Thermo Scientific Supplier No. 371370010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-98-3 | C8H5F | 120.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-98-3
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.13
MDL Number MFCD00168823
InChI Key QXSWHQGIEKUBAS-UHFFFAOYSA-N
Synonym 4-fluorophenylacetylene, benzene, 1-ethynyl-4-fluoro, 1-ethynyl-4-fluoro-benzene, 4'-fluorophenyl acetylene, p-fluorophenyl acetylene, 4'-fluorophenylacetylene, pubchem16719, 4-fluorophenyl acetylene, 4-fluoro phenylacetylene
PubChem CID 522627
IUPAC Name 1-ethynyl-4-fluorobenzene
SMILES C#CC1=CC=C(C=C1)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 27.0°C to 28.0°C
Color Colorless to Yellow
Density 1.0480g/mL
Boiling Point 55.0°C to 56.0°C (40.0 mmHg)
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4C≡CH
Refractive Index 1.5150 to 1.5170
Quantity 1 g
Specific Gravity 1.048
Formula Weight 120.13
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 1-Ethynyl-4-fluorobenzene, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethynyl-4-fluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.