missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-2-fluorobenzene, 97%, Thermo Scientific Chemicals

Product Code. p-3211397 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10609224

Brand: Thermo Scientific Acros 380992500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-49-4
Molecular Formula C8H5F
Molecular Weight (g/mol) 120.13
MDL Number MFCD00799540
InChI Key YFPQIXUNBPQKQR-UHFFFAOYSA-N
Synonym 2-fluorophenylacetylene, 2'-fluorophenyl acetylene, 1-ethynyl-2-fluoro-benzene, 2'-fluorophenylacetylene, 2-fluorophenylethyne, o-fluorophenylacetylene, benzene, ethynylfluoro, 2-fluoro-phenylacetylene, 2-fluorophenyl-ethyne, 2-fluorophenyl acetylene
PubChem CID 643358
IUPAC Name 1-ethynyl-2-fluorobenzene
SMILES FC1=CC=CC=C1C#C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.0600g/mL
Boiling Point 150°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H4C∽CH
Refractive Index 1.5240 to 1.527
Quantity 250 mg
Specific Gravity 1.06
Formula Weight 120.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Ethynyl-2-fluorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethynyl-2-fluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.