missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-1-cyclohexanol, 99+%

Product Code. 10326510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10326510 500 g Glass bottle 500g
10569600 100 g Glass bottle 100g
11974641 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10326510 Supplier Thermo Scientific Supplier No. 119105000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-27-3 | C8H12O | 124.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-27-3
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00003858
InChI Key QYLFHLNFIHBCPR-UHFFFAOYSA-N
Synonym 1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol
PubChem CID 6525
IUPAC Name 1-ethynylcyclohexan-1-ol
SMILES C#CC1(CCCCC1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 30.0°C to 32.0°C
Color Colorless to Yellow
Density 0.9763g/mL
Boiling Point 180.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CC6H10OH
Refractive Index 1.4810 to 1.4840
Quantity 500 g
Beilstein 06,II,1
Merck Index 10,381
Specific Gravity 0.9763
Solubility Information Solubility in water: 24.5g/L (20°C). Other solubilities: soluble in most organic solvents, soluble in diethyl ether, alcohols, esters, ketone, and chlorinated hydrocarbons
Formula Weight 124.18
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material 1-Ethynyl-1-cyclohexanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Ethynyl-1-cyclohexanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.