missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpyridinium bromide, 99%

Product Code. 11449026
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449026 25 g 25g
11459026 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449026 Supplier Thermo Scientific Alfa Aesar Supplier No. A17911.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1906-79-2 | C7H10BrN | 188.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1906-79-2
Molecular Formula C7H10BrN
Molecular Weight (g/mol) 188.068
MDL Number MFCD00041999
InChI Key ABFDKXBSQCTIKH-UHFFFAOYSA-M
Synonym 1-ethylpyridinium bromide, 1-ethylpyridin-1-ium bromide, ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide, n-ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide 1:1, pyridineethobromide, ethylpyridiniumbromide, 1-ethyl-pyridiniubromide, n-ethyl pyridinium bromide
PubChem CID 15927
IUPAC Name 1-ethylpyridin-1-ium;bromide
SMILES CC[N+]1=CC=CC=C1.[Br-]

Specifications

Melting Point 117°C to 121°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 188.07
Percent Purity 99%
Chemical Name or Material 1-Ethylpyridinium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.