missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylcyclohexanol, 97%

Product Code. p-4812609 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 15279856

Brand: Thermo Scientific Acros 453350050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1940-18-7 | C8H16O | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1940-18-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00021402
InChI Key BUCJHJXFXUZJHL-UHFFFAOYSA-N
Synonym 1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
PubChem CID 16021
IUPAC Name 1-ethylcyclohexan-1-ol
SMILES CCC1(O)CCCCC1

Specifications

Density 0.9190g/mL
Boiling Point 166.0°C
Flash Point 68°C
Assay Percent Range 96% min. (GC)
Quantity 5 mL
Specific Gravity 0.919
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 1-Ethylcyclohexanol
compliance-icons
Product Identifier
  • 1-Ethylcyclohexanol, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.