missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Mikromol™
Description
1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid |
| CAS | 70459-04-0 |
| Analyte or Component Names | 1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 F N3 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Norfloxacin Imp. G (EP), Norfloxacin Impurity G |
| InChI Formula | InChI=1S/C17H18FN3O4/c1-2-20-9-12(17(24)25)16(23)11-7-13(18)15(8-14(11)20)21-5-3-19(10-22)4-6-21/h7-10H,2-6H2,1H3,(H,24,25) |
| SMILES | CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(CC3)C=O |
| IUPAC Name | 1-ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?