missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-eth-1-ynylbenzene, Thermo Scientific™

Product Code. 10401864
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10401864 25 g 25g
10752843 1 g 1g
10181292 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10401864 Supplier Thermo Scientific Maybridge Supplier No. BTB09906EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40307-11-7
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00173887
InChI Key ZNTJVJSUNSUMPP-UHFFFAOYSA-N
Synonym 4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
PubChem CID 142425
IUPAC Name 1-ethyl-4-ethynylbenzene
SMILES CCC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 0.93g/mL
Assay Percent Range 97%
Linear Formula HC2CC6H4CH2CH3
Quantity 25 g
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 1-Ethyl-4-eth-1-ynylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.