missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene, 99+%, Thermo Scientific™

Product Code. 11421470
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421470 10 g 10g
11411470 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11421470 Supplier Thermo Scientific Alfa Aesar Supplier No. H26908.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117923-34-9
Molecular Formula C22H26
Molecular Weight (g/mol) 290.45
MDL Number MFCD09909569
InChI Key ZCEJWGXSGOFJTA-UHFFFAOYSA-N
Synonym 1-ethyl-4-4-n-hexylphenyl ethynyl benzene, 1-ethyl-4-2-4-n-hexylphenyl ethynyl benzene, 1-ethyl-4-2-4-hexylphenyl ethynyl benzene, 1-2-4-ethylphenyl ethynyl-4-hexylbenzene, acmc-20aov8, 4-ethyl-4'-n-hexyldiphenylacetylene, 1-ethyl-4-4-hexylphenyl ethynyl benzene, 4-4-ethylphenyl ethynyl-1-hexylbenzene
PubChem CID 18952851
IUPAC Name 1-ethyl-4-[2-(4-hexylphenyl)ethynyl]benzene
SMILES CCCCCCC1=CC=C(C=C1)C#CC1=CC=C(CC)C=C1

Specifications

Melting Point 10°C to 12°C
Boiling Point 172°C to 174°C (0.5 mmHg)
Refractive Index 1.59
Quantity 10 g
Formula Weight 290.45
Percent Purity ≥99%
Chemical Name or Material 1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.