missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium thiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 15495828
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495828 5 g 5g
15405838 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495828 Supplier Thermo Scientific Alfa Aesar Supplier No. H59493.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 331717-63-6 | Molecular Formula: C7H11N3S | Molecular Weight: 169.246 g/mol

It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and also used as primary and secondary intermediate.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331717-63-6
Molecular Formula C7H11N3S
Molecular Weight (g/mol) 169.246
MDL Number MFCD06798182
InChI Key VASPYXGQVWPGAB-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium thiocyanate, basionics™ vs 01, dsstox_cid_27860, dsstox_rid_82613, dsstox_gsid_47884, 1-methyl-3-ethylimidazolium thiocyanate, 1-ethyl-3-methyl imidazolium thio-cyanate, 1-ethyl-3-methyl-1h-imidazol-3-ium thiocyanate, 1-ethyl-3-methylimidazolium thiocyanate h-nmr, hplc, 1-ethyl-3-methylimidazolium thiocyanate, for electrochemistry t
PubChem CID 16211115
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;thiocyanate
SMILES CCN1C=C[N+](=C1)C.C(#N)[S-]

Specifications

Melting Point -6°C
Density 1.14
Refractive Index n20/D 1.553
Quantity 5 g
UN Number UN2810
Sensitivity Moisture sensitive
Formula Weight 169.25
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium thiocyanate
compliance-icons
Product Identifier
  • 1-Ethyl-3-methylimidazolium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-Ethyl-3-methylimidazolium thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.