missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%

Product Code. 11349705
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349705 2 g 2g
11359705 10 g 10g
11369705 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349705 Supplier Thermo Scientific Alfa Aesar Supplier No. L19762.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 155371-19-0 | C6H11F6N2P | 256.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155371-19-0
Molecular Formula C6H11F6N2P
Molecular Weight (g/mol) 256.132
MDL Number MFCD00216703
InChI Key DPDAKOVGQUGTHH-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc
PubChem CID 2758873
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate
SMILES CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 58°C to 62°C
Quantity 2 g
Sensitivity Hygroscopic
Formula Weight 256.13
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.