missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium ethyl sulfate, 99%

Product Code. 11446460
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446460 250 g 250g
11436460 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446460 Supplier Thermo Scientific Alfa Aesar Supplier No. H27571.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 342573-75-5 | C8H16N2O4S | 236.286 g/mol

1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.

Solubility
Miscible with water and alcohol.

Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 342573-75-5
Molecular Formula C8H16N2O4S
Molecular Weight (g/mol) 236.286
MDL Number MFCD06798189
InChI Key VRFOKYHDLYBVAL-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate
PubChem CID 12095229
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;ethyl sulfate
SMILES CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-]

Specifications

Melting Point <-30°C
Density 1.24
Flash Point 162°C (324°F)
Quantity 250 g
Solubility Information Miscible with water and alcohol.
Formula Weight 236.29
Percent Purity 99%
Chemical Name or Material 1-Ethyl-3-methylimidazolium ethyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.