missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium diethyl phosphate, 98%

Product Code. 15473677
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473677 25 g 25g
11357209 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473677 Supplier Thermo Scientific Alfa Aesar Supplier No. H27400.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 848641-69-0 | C10H21N2O4P | 264.26 g/mol

It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 848641-69-0
Molecular Formula C10H21N2O4P
Molecular Weight (g/mol) 264.26
MDL Number MFCD09953486
InChI Key HQWOEDCLDNFWEV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc
PubChem CID 53316406
IUPAC Name diethyl phosphate;1-ethyl-3-methylimidazol-3-ium
SMILES CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC

Specifications

Melting Point 19°C to 21°C
Density 1.157
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 264.26
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium diethyl phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.