missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium dicyanamide, 98%

Product Code. 11481450
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481450 5 g 5g
11491450 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481450 Supplier Thermo Scientific Alfa Aesar Supplier No. H26901.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 370865-89-7 | C8H11N5 | 177.211 g/mol

Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Soluble in water. Soluble in solvents like methanol, acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 370865-89-7
Molecular Formula C8H11N5
Molecular Weight (g/mol) 177.211
MDL Number MFCD08276373
InChI Key MKHFCTXNDRMIDR-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium dicyanamide, ethylmethylimidazolium dicyanamide, emim dca, acmc-209iqn, basionics™ vs 03, dsstox_cid_27940, dsstox_rid_82692, dsstox_gsid_47964, ksc221g0h
PubChem CID 11159638
IUPAC Name cyanoiminomethylideneazanide;1-ethyl-3-methylimidazol-3-ium
SMILES CCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Quantity 5 g
Solubility Information Soluble in water. Soluble in solvents like methanol,acetone.
Formula Weight 177.21
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium dicyanamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.