missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium chloride, 97%

Product Code. p-3736474 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10362873

Brand: Thermo Scientific Acros 354080050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65039-09-0
Molecular Formula C6H11ClN2
Molecular Weight (g/mol) 146.62
MDL Number MFCD00074843
InChI Key BMQZYMYBQZGEEY-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180
PubChem CID 2734160
ChEBI CHEBI:61327
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;chloride
SMILES [Cl-].CCN1C=C[N+](C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77.0°C to 79.0°C
Color Yellow
Flash Point 186°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Viscosity 61.2 mPa.s (80°C)
Formula Weight 146.62
Percent Purity 97%
Physical Form Agglomerating Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.