missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-bromoadamantane, TRC

Product Code. 30387233
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387233 1 g -
30377426 250 mg -
30377865 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387233 Supplier Toronto Research Chemicals Supplier No. TRCE9002801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878-61-5
Molecular Formula C12H19Br
Molecular Weight (g/mol) 243.18
Synonym 1-Bromo-3-ethyladamantane, 1-Bromo-3-ethyltricyclo[3.3.1.13,7]decane
SMILES CC[C@@]12C[C@H]3C[C@H](C[C@@](Br)(C3)C1)C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H19Br/c1-2-11-4-9-3-10(5-11)7-12(13,6-9)8-11/h9-10H,2-8H2,1H3
Formula Weight 242.067
Chemical Name or Material 1-Ethyl-3-bromoadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.