missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea, Mikromol™

Product Code. 17951276
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951276 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951276 Supplier Mikromol Supplier No. MM1283.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea
CAS 58155-35-4
Analyte or Component Names 1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea
Shipping Condition Room Temperature
Molecular Formula C15 H18 N4 O3 S
Certifications/Compliance ISO 17025
Synonym Torasemide Imp. C (EP), Torsemide USP Related Compound C, Torsemide USP RC C, 1-Ethyl-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea, Torasemide, Anhydrous Imp. C (EP), Torsemide Related Compound C, Torasemide, Anhydrous Impurity C, Torasemide Impurity C
InChI Formula InChI=1S/C15H18N4O3S/c1-3-17-15(20)19-23(21,22)14-10-16-8-7-13(14)18-12-6-4-5-11(2)9-12/h4-10H,3H2,1-2H3,(H,16,18)(H2,17,19,20)
SMILES CCNC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2
IUPAC Name 1-ethyl-3-[4-(3-methylanilino)pyridin-3-yl]sulfonylurea
Molecular Weight (g/mol) 334.39
Quantity 50 mg
Formula Weight 334.11 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.