missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals
25.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
Solubility
Soluble in Water.
Notes
Precursor in synthesis of carbocyanine dyes.
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.
Solubility
Soluble in Water.
Notes
Precursor in synthesis of carbocyanine dyes.
Specifications
Melting Point | 231°C (decomposition) |
Quantity | 5 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in Water. |
Formula Weight | 299.16 |
Percent Purity | 97% |
Chemical Name or Material | 1-Ethyl-2-methylquinolinium iodide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'