missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2-methylquinolinium iodide, 97%

Product Code. 11483244
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483244 25 g 25g
11473244 5 g 5g
11493244 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483244 Supplier Thermo Scientific Alfa Aesar Supplier No. A10584.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 606-55-3 | C12H14IN | 299.155 g/mol

1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

Solubility
Soluble in Water.

Notes
Precursor in synthesis of carbocyanine dyes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-55-3
Molecular Formula C12H14IN
Molecular Weight (g/mol) 299.155
MDL Number MFCD00031829
InChI Key OEVSHJVOKFWBJY-UHFFFAOYSA-M
Synonym 1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
PubChem CID 69076
IUPAC Name 1-ethyl-2-methylquinolin-1-ium;iodide
SMILES CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]

Specifications

Melting Point 231°C (decomposition)
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Soluble in Water.
Formula Weight 299.16
Percent Purity 97%
Chemical Name or Material 1-Ethyl-2-methylquinolinium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.