missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium chloride, 97%, Thermo Scientific Chemicals

Product Code. 15465468
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used reaction media for chemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used reaction media for chemical processes.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92507-97-6
Molecular Formula C7H13N2
Molecular Weight (g/mol) 125.19
MDL Number MFCD03093290
InChI Key IRGDPGYNHSIIJJ-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium chloride, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium chloride, ksc496m4d, 1,2-dimethyl-3-ethylimidazolium chloride, 1-ethyl-2,3-dimethyl-imidazolium chloride, 1-ethyl-2,3-dimethylimidazol-3-ium chloride, 1-ethyl-2,3-dimethylimidazol-1-ium chloride, 3-ethyl-1,2-dimethylimidazol-1-ium chloride, 1-ethyl-2,3-dimethylimidazolium chloride t
PubChem CID 2734165
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;chloride
SMILES CCN1C=C[N+](C)=C1C

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 160.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'