missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-2,3-dimethylimidazolium chloride, 97%
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
Description
It is used reaction media for chemical processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used reaction media for chemical processes.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is used reaction media for chemical processes.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Quantity | 5 g |
| Sensitivity | Moisture sensitive |
| Formula Weight | 160.64 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | 1-Ethyl-2,3-dimethylimidazolium chloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction