missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-1,2-dihydro-2,2,4-trimethyl-7-quinolinol Hydrochloride, TRC

Product Code. 30385110
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385110 500 mg 500mg
30373540 1 g 1g
30372436 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385110 Supplier Toronto Research Chemicals Supplier No. TRCE678520500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Ethyl-1,2-dihydro-2,2,4-trimethyl-7-quinolinol Hydrochloride
Analyte or Component Names 1-Ethyl-1,2-dihydro-2,2,4-trimethyl-7-quinolinol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H19 N O . H Cl
Synonym N-Ethyl-7-hydroxy-2,2,4-trimethyl-1,2-dihydroquinoline Hydrochloride
InChI Formula InChI=1S/C14H19NO.ClH/c1-5-15-13-8-11(16)6-7-12(13)10(2)9-14(15,3)4;/h6-9,16H,5H2,1-4H3;1H
Solution Type Neat
SMILES Cl.CCN1c2cc(O)ccc2C(=CC1(C)C)C
IUPAC Name 1-ethyl-2,2,4-trimethylquinolin-7-ol;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 253.768
Quantity 500 mg
Formula Weight 253.123
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.