missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxyethyl-indacaterol, TRC

Product Code. 30392675
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392675 5 mg 5mg
30391891 50 mg -
30401281 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392675 Supplier Toronto Research Chemicals Supplier No. TRCI4997735MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2250243-41-3
Molecular Formula C26H32N2O3
Molecular Weight (g/mol) 420.55
Synonym 5-[2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-ethoxyethyl]-8-hydroxy-2(1H)-quinolinone, Indacaterol Impurity 3
IUPAC Name 5-[2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-ethoxyethyl]-8-hydroxy-1H-quinolin-2-one
SMILES CCOC(CNC1Cc2cc(CC)c(CC)cc2C1)c3ccc(O)c4NC(=O)C=Cc34

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H32N2O3/c1-4-16-11-18-13-20(14-19(18)12-17(16)5-2)27-15-24(31-6-3)21-7-9-23(29)26-22(21)8-10-25(30)28-26/h7-12,20,24,27,29H,4-6,13-15H2,1-3H3,(H,28,30)
Formula Weight 420.2413
Chemical Name or Material 1-Ethoxyethyl-indacaterol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.