missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxyethanimine-D3 Hydrochloride, TRC

Product Code. 30381271
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381271 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381271 Supplier Toronto Research Chemicals Supplier No. TRCE749290500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Ethoxyethanimine-D3 Hydrochloride
CAS 74929-87-6
Molecular Formula C4H6D3NO . HCl
Quantity 500 mg
Synonym Ethyl Acetimidate HydrochlorideEthyl Acetoamidate HydrochlorideEthyl Acetocarboximidate HydrochlorideEthyl Acetoimidate HydrochlorideEthyl Ethanimidate HydrochlorideEthyl Ethanimidoate HydrochlorideEthyl Iminoacetate hydrochloride
InChI Formula InChI=1S/C4H9NO.ClH/c1-3-6-4(2)5;/h5H,3H2,1-2H3;1H/i2D3;
SMILES Cl.[2H]C([2H])([2H])C(=N)OCC
IUPAC Name ethyl 2,2,2-trideuterioethanimidate;hydrochloride
Molecular Weight (g/mol) 126.6
Formula Weight 126.0639
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.