missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide, 97%

Product Code. 11429943
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429943 25 g 25g
11439943 100 g 100g
11449943 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429943 Supplier Thermo Scientific Alfa Aesar Supplier No. A13023.14
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 30018-16-7 | C23H24BrO2P | 443.32 g/mol

1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30018-16-7
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.32
MDL Number MFCD00031605
InChI Key RSYXORMKBUFAMS-UHFFFAOYNA-M
Synonym 1-ethoxy-1-oxopropan-2-yl triphenylphosphonium bromide, 1-ethoxycarbonyl ethyltriphenylphosphonium bromide, 1-ethoxycarbonyl ethyl triphenylphosphonium bromide, 2-ethoxy-1-methyl-2-oxoethyl triphenylphosphonium bromide, ceetppb, 1-carbethoxyethyl triphenylphosphonium bromide, 1-ethoxycarbonylethyl triphenylphosphonium bromide, 1-ethoxy-1-oxopropan-2-yl triphenylphosphanium bromide, phosphonium, 2-ethoxy-1-methyl-2-oxoethyl triphenyl-, bromide, ethoxycarbonylethyltriphenylphosphoniumbromide
PubChem CID 3084511
IUPAC Name (1-ethoxy-1-oxopropan-2-yl)-triphenylphosphanium;bromide
SMILES [Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼130°C (decomposition)
Quantity 25 g
Beilstein 3822244
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 443.33
Percent Purity 97%
Chemical Name or Material (1-Ethoxycarbonylethyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.