missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol, 90+%, remainder 2-ethoxy-1-propanol

Product Code. 11490560
Change view
Click to view available options
Quantity:
500 mL
4 x 500 mL
Unit Size:
2000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490560 4 x 500 mL 2000mL
11480560 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490560 Supplier Thermo Scientific Alfa Aesar Supplier No. 043306.AY
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol

1-Ethoxy-2-propanol is used as a solvent for a wide variety of resins such as epoxies, acrylic, alkyds, polyesters, nitrocellulose, and polyurethanes. It is also used in the surface coating and printing industries, thereby it regulate the flow, levelling, and coalescence of both surface coatings. It acts as an intermediate in the production of agro-chemicals and de-icing, and anti-icing formulations. It finds application as to make cleaning products, as grease and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1569-02-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00067050
InChI Key JOLQKTGDSGKSKJ-UHFFFAOYNA-N
Synonym 1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
PubChem CID 15287
IUPAC Name 1-ethoxypropan-2-ol
SMILES CCOCC(C)O

Specifications

Melting Point -100°C
Color Colorless
Density 0.896 g/mL
Boiling Point 132°C
Flash Point 42°C (108°F)
Linear Formula CH3CH2OCH2CH(OH)CH3
Refractive Index 1.4065
Quantity 4 x 500 mL
UN Number UN1987
Solubility Information Miscible with water.
Formula Weight 104.15
Percent Purity ≥90%
Physical Form Liquid
Assay Remainder 2-ethoxy-1-propanol
Chemical Name or Material 1-Ethoxy-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.