missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanethiol, 98%

Product Code. 15497447
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497447 250 mg 250mg
15487447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497447 Supplier Thermo Scientific Alfa Aesar Supplier No. H34498.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13373-97-2 | C20H42S | 314.616 g/mol

1-Eicosanethiol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.

Notes
Air and moisture sensitive. Incompatible with heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13373-97-2
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.616
MDL Number MFCD07779406
InChI Key YYHYWOPDNMFEAV-UHFFFAOYSA-N
Synonym 1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
PubChem CID 139443
IUPAC Name icosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCS

Specifications

Melting Point 38°C to 41°C
Odor Stench
Linear Formula CH3(CH2)19SH
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 314.61
Percent Purity 98%
Chemical Name or Material 1-Eicosanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.